1,1,1-Trifluoro-2-butanone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

FL1574.5 FL1574.25 001574-5G
CHMPFL1574.5EA 0 GBP
CHMPFL1574.5 CHMPFL1574.25 FLUO001574-5G
1,1,1-Trifluoro-2-butanone
1,1,1-Trifluoro-2-butanone
Formula: C₄H₅F₃O
MDL Number: MFCD00190641
CAS Number: 381-88-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR