3-Fluorocinnamic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 3-(3-Fluorophenyl)acrylic acid, m-Fluorocinnamic acid, 3-Fluorocinnamic Acid

001441-25G 001441-100G
FLUO001441-25GEA 0 GBP
FLUO001441-25G FLUO001441-100G
3-Fluorocinnamic acid
3-Fluorocinnamic acid
Formula: C₉H₇FO₂
MW: 166.15 g/mol
MDL Number: MFCD00004383
CAS Number: 458-46-8
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR