Isopropyl trifluoroacetate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

001203.100G 001203-500G
FLUO001203.100GEA 0 GBP
FLUO001203.100G FLUO001203-500G
Isopropyl trifluoroacetate
Isopropyl trifluoroacetate
Formula: C₅H₇F₃O₂
MDL Number: MFCD00013566
CAS Number: 400-38-4
EINECS: 206-922-1
UN: 2924
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR