1-Bromo-2-fluorobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

001121-100G 001121-500G 001121-25G
FLUO001121-100GEA 0 GBP
FLUO001121-100G FLUO001121-500G FLUO001121-25G
1-Bromo-2-fluorobenzene
1-Bromo-2-fluorobenzene
Formula: C₆H₄BrF
Boiling Pt: 155 °C (1013 hPa)
Density: 1.61 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000282
CAS Number: 1072-85-1
EINECS: 214-018-3
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR