Vacuum wand, F-series
Supplier: WINDRUSH TECHNOLOGY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
F003-Y-14-3F F003-X-06-3F F003-X-06-PK F003-Z-05-3F F003-Z-05-PK F003-Z-05-VP
FLUMF003-Y-14-3FEA 0 GBP
FLUMF003-Y-14-3F FLUMF003-X-06-3F FLUMF003-X-06-PK FLUMF003-Z-05-3F FLUMF003-Z-05-PK FLUMF003-Z-05-VP
Vacuum wand, F-series
Vacuum Handling Systems
This F series wand bodies are manufactured in teflon and are the cleanest means of wafer handling. Tips are available in PCTFE, PEEK, Vespel and stainless steel. The tips are optimally sized for each wafer diameter, ensuring that contact is both minimal and secure.
  • Polished contact surface provides excellent adhesion to water
  • Fixed tip, collect joint and swivel joint are three body styles
  • Unique control valve ensures reliable suction and instantaneous release with out particle genertion