Restriction enzymes, EcoRI, FastDigest™, Fermentas

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

FD0274 FD0275
FD0274EA 70.2 GBP
FD0274 FD0275
Restriction enzymes, EcoRI, FastDigest™, Fermentas
Nucleic Acid Reagents Restriction Enzymes

Thermo Scientific™ FastDigest™ enzymes are an advanced line of restriction enzymes for rapid DNA digestion.


  • Activity (100%) of all FastDigest™ enzymes in one universal reaction buffer
  • Direct loading on gels with FastDigest™ Green buffer
  • Buffer compatibility (100%) with downstream applications


Designed to complete digestion in 5 to 15 minutes.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR