Avantor® ACE®, UHPLC Pre-Column Filters
Supplier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
EXL-PCF10/ACQEA 656 GBP
EXL-PCF10/ACQ EXL-PCF10 EXL-PCF05/ACQ EXL-PCF05 EXL-PCF01/ACQ EXL-PCF01
Avantor® ACE®, UHPLC Pre-Column Filters
Filters Precolumn Filters and Frits
Avantor® ACE® UHPLC pre-column filters made from stainless steel/PEEK provide protection for all UHPLC columns. They offer ultra-low dispersion for maximum compatibility with UHPLC instruments and columns. For use with Waters 'Classic' Acquity® systems, an alternative Waters Acquity® compatible version provides improved performance.
  • Universal filter, suitable for all UHPLC columns
  • Compatible with all UHPLC systems
  • Pressure rated to 1000 bar
  • Reusable