Hand sanitiser gel
Supplier: Deb-STOKO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ISG1L
EVONISG1LEA 0 GBP
EVONISG1L
Hand sanitiser gel
Skincare Skin Creams Hand and Body Lotions
Deb stoko sanitise gel dispenser can be used in office and facility, industrial, food, hospitality and education sectors.
  • Glove compatible - can be applied to hands prior to wearing latex, vinyl and nitrile gloves
  • No water required - quickly dries on hands without the need for rinsing with water
Deb® InstantGel advantages include highly effective formula is fully tested and proven to be bactericidal, virucidal, yeasticidal and mycobactericidal. Kills 99,999% of many common germs, rapidly and easily rubs across hands, providing the user with completeMore Product Information