Anti-PTGR2 Goat Polyclonal Antibody

Supplier: Everest Biotech
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

EB12931
EVBIEB12931EA 330 GBP
EVBIEB12931
Anti-PTGR2 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PTGR2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Pig
Order Now

SPECIFICATIONS

Antigen symbol PTGR2
Environmentally Preferable
Antigen synonyms PTGR2|zinc-binding alcohol dehydrogenase domain-containing protein 1|15-oxoprostaglandin-delta13-reductase|ZADH1|PGR2|PRG-2|domain containing 1|15-oxoprostaglandin 13-reductase|prostaglandin reductase 2|zinc binding alcohol dehydrogenase
Antibody type Primary
Clonality Polyclonal
Reactivity Pig,Human
Host Goat
Gene ID 145482
Isotype IgG
Western blot Yes
ELISA Yes
Immunogen PTGR2 / ZADH1
Sequence C-PDNINEGQVQVRT
Format Affinity Purified
Form Liquid
Molecular weight 38.5
Concentration 0,5 µg/µl
Storage buffer Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.
Storage temperature +8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR