Anti-PTGES2 Goat Polyclonal Antibody

Supplier: Everest Biotech
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

EB12928
EVBIEB12928EA 330 GBP
EVBIEB12928
Anti-PTGES2 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PTGES2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR