Anti-SON Goat Polyclonal Antibody

Supplier: Everest Biotech
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

EB10825
EVBIEB10825EA 330 GBP
EVBIEB10825
Anti-SON Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SON (SON DNA binding protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR