Anti-Calcipressin-1 Goat polyclonal antibody unconjugated

Supplier: Everest Biotech
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

EB10693
EVBIEB10693EA 330 GBP
EVBIEB10693
Anti-Calcipressin-1 Goat polyclonal antibody unconjugated
Antibodies
Type: Primary
Antigen: Calcipressin-1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR