Anti-KPNB1 Goat Polyclonal Antibody

Supplier: Everest Biotech
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

EB07018
EVBIEB07018EA 330 GBP
EVBIEB07018
Anti-KPNB1 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: KPNB1 (karyopherin (importin) beta 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR