Conductivity meter, handheld, Eutech CyberScan COND 600
Supplier: EUTECH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ECCONWP60043K
EUTEECCONWP60043KEA 1630 GBP
EUTEECCONWP60043K
Conductivity meter, handheld, Eutech CyberScan COND 600
Electrochemistry Meters Conductivity Meters
The waterproof, portable conductivity/TDS meter features a broad conductivity range with auto-ranging capabilities for more convenience.
  • Mega-memory space that stores/ recalls up to 500 data sets formatted to meet GLP standards
  • Wireless and cable-free data transfer with infrared IrDA technology.
  • Large, easy-to-read custom screen with simultaneous information display allows optimum viewing with user-friendly icons
  • HOLD function, auto-power off feature
  • Self-diagnostic with message codes for easy troubleshooting
  • Rugged and waterproof to IP67 standard with
More Product Information