Ductless filtering chemical storage cabinets, Captair® Smart
Supplier: ERLAB CAPTAIRE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
AB46A0000332 AB46A0000432 AB56A000030332 AB56A000040432
ERLAAB46A0000332EA 3620 GBP
ERLAAB46A0000332 ERLAAB46A0000432 ERLAAB56A000030332 ERLAAB56A000040432
Ductless filtering chemical storage cabinets, Captair® Smart
Fume Hoods and Enclosures Ductless Fume Hoods
The Captair® Smart filtering chemical storage cabinets are designed with a simple and innovative way of communication. Smart Technology uses simple light to show that the unit is operating safely so the user can focus his attention on what is most important: his work. Smart Technology enables visual light to indicate the hood status. A simple light pattern indicates if the air flow is compromised, if there is a fan failure, a filter breakthrough or if the doors are open.
Please complete the quote request to enable us to assist you in finding the right unit for your application.
https://uk.cmd2.More Product Information