Restriction enzymes, Eco32I (EcoRV), Fermentas
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ER0301 ER0302 ER0305
ER0301EA 61.2 GBP
ER0301 ER0302 ER0305
Restriction enzymes, Eco32I (EcoRV), Fermentas
Nucleic Acid Reagents Restriction Enzymes
Eco32I (EcoRV) restriction enzyme recognizes GAT^ATC sites and cuts best at 37 °C in R buffer Isoschizomers: EcoRV.
  • Superior quality—stringent quality control and industry leading manufacturing process
  • Convenient color-coded Five Buffer System
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities
Used in molecular cloning, restriction site mapping, genotyping, southern blotting and restriction fragment length polymorphism (RFLP).