RackScan
Supplier: EPPENDORF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
6002000.006 6002000.014
EPPE6002000.006EA 9330 GBP
EPPE6002000.006 EPPE6002000.014
RackScan
Barcode and RFID Systems
The RackScan instrument can read up to 96 barcoded vials in parallel.
  • Camera based instrument resulting in rapid imaging and decoding
  • Comfortable and easy plug-in system
  • Small footprint, ideal for benchtop (and integration)
The RackScan can be equipped with the 's' accessory.