Centrifuges, benchtop, ventilated/refrigerated, 5810 / 5810 R (General Lab Product)
Supplier: EPPENDORF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5810000.060 5810000.365 5810000.460 5810000.560 5810000.665 5811000.065 5811000.360 5811000.465 5811000.660 5811000.865
EPPE5810000.060EA 5000 GBP
EPPE5810000.060 EPPE5810000.365 EPPE5810000.460 521-2793 EPPE5810000.665 EPPE5811000.065 EPPE5811000.360 EPPE5811000.465 EPPE5811000.660 EPPE5811000.865
Centrifuges, benchtop, ventilated/refrigerated, 5810 / 5810 R (General Lab Product)
Centrifuges Benchtop Centrifuges
This reliable centrifuge for medium to high-throughput applications combines extraordinary versatility and capacity for both tubes and plates with an extraordinary compact footprint. Bucket adapters and adapter modules can be stacked until the required tube height is attained and then lowered onto the holders in the rectangular buckets. The stable plastic adapters are robustly constructed and can be easily cleaned, even autoclaved in their entirety in the event of contamination with biologically active sample material.
  • Automatic rotor recognition senses rotor type and sets maximum
More Product Information