Anti-RASAL2 Rabbit Polyclonal Antibody

Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PSC-15-887-R05
ENZOPSC-15-887-R05EA 0 GBP
ENZOPSC-15-887-R05
Anti-RASAL2 Rabbit Polyclonal Antibody
Antibodies
Caution: For research use only.

Type:
Antigen: RASAL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR