Insulin ELISA kit
Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ENZKIT1410001
ENZOENZKIT1410001EA 516 GBP
ENZOENZKIT1410001
Insulin ELISA kit
Assays ELISAs
Highly sensitive Insulin ELISA kit enabling detection of insulin in serum, plasma, tissue culture media in just 3 hours.
  • Highly sensitive measurement of insulin, detecting as little as 21,6 pg/ml (0,622 µIU/ml)
  • Negligible cross reactivity with proinsulin
  • High throughput format with results in 3 hours for up to 40 samples in duplicate
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of insulin in serum, plasma, and tissue culture media. The Insulin ELISA kit is a colorimetric immunometric immunoassay kit with results in 3 hours.
Ordering information: Contains Microtiter plate, Assay Buffer 13, Standard, Wash Buffer Concentrate, Antibody, Antibody Diluent, Conjugate, Substrate, and Stop Solution.