E. coli host cell protein ELISA kit
Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ENZKIT1270001
ENZOENZKIT1270001EA 1060 GBP
ENZOENZKIT1270001
E. coli host cell protein ELISA kit
Assays ELISAs
Sensitive (30 ng/ml) ELISA for the quantitative determination of host cell protein contamination in bulk products expressed in E. coli expression systems.
For the quantitative determination of contamination with E.coli host cell protein.
The E. coli host cell protein ELISA kit is a complete, colorimetric, immunometric immunoassay kit for the quantitative determination of E. coli host cell protein contamination with results in 3 hours
Ordering information: Contains 1 x 96 well plate, protein standards, 5x dilution buffer, 10x PBS-T, reporting antibody, streptavidin-HRP conjugate, TMB substrate, stop solution, plate sealer.