AGK7 99 HPLC_ASSAY_METHOD

Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ENZCHM1460005 ENZCHM1460010
ENZOENZCHM1460005EA 144 GBP
ENZOENZCHM1460005 ENZOENZCHM1460010
AGK7 99 HPLC_ASSAY_METHOD
AGK7

Selectively inhibits sirtuin 3 over sirtuins 1 and 2.


Can be used as an inactive control in experiments with AGK2.

MW: 434.3 g/mol
CAS Number: 304896-21-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=953e981d46ba7ada3ae4cafc9a07e153f3bc2554&vid=3515e5f59ca9a8bec212f7035b2959db486f4529&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR