Anti-CXCL11 Mouse Monoclonal Antibody (Biotin) [clone: 22F1]

Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ENZABS2750100
ENZOENZABS2750100EA 516 GBP
ENZOENZABS2750100
Anti-CXCL11 Mouse Monoclonal Antibody (Biotin) [clone: 22F1]
Antibodies
Recommended Applications: ELISA, WB

Species reactivity: Human

Type: Primary
Antigen: CXCL11
Clonality: Monoclonal
Clone: 22F1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR