Anti-Biotin Rabbit Polyclonal Antibody

Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ENZ328936000
ENZOENZ328936000EA 543 GBP
ENZOENZ328936000
Anti-Biotin Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Biotin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Species independent
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR