SUMO-1 (93-97)-AMCA
Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BMLUW05001000
ENZOBMLUW05001000EA 165 GBP
ENZOBMLUW05001000
SUMO-1 (93-97)-AMCA
Proteins and Peptides
SUMO1-AMCA is a synthetic AMCA-peptide substrate based on the C-terminal residues of SUMO1 and is a useful fluorogenic substrate for deSUMOylating enzymes, including SENP1 (BML-UW9760).
  • Provides an alternative substrate to SUMO1-AMC
It has particular utility for the study of deSUMOylating activity where continuous monitoring of activity is essential.