Haloenol lactone suicide substrate 98 TLC_ASSAY_METHOD

Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: HELSS

BMLST3400005 BMLST3400025
ENZOBMLST3400005EA 162 GBP
ENZOBMLST3400005 ENZOBMLST3400025
Haloenol lactone suicide substrate 98 TLC_ASSAY_METHOD
Haloenol lactone suicide substrate

HELSS is a potent, irreversible, mechanism-based inhibitor of Ca2+-independent PLA2 (70% inhibition at 100 nM). It possesses a 1000-fold selectivity for Ca2+-independent PLA2 vs. Ca2+-dependent PLA2 . It has been used to identify the PLA2 involved in the release of arachidonate induced by vasopressin stimulation of A-10 smooth muscle cells (IC50=1 µM). It has also been shown to inhibit the Mg2+-dependent phophatidate phosphohydrolase from P388D1 macrophages (IC50~8 µM).


  • Phospholipase inhibitor


White solid.


Soluble in DMSO (25mg/ml).

Formula: C₁₆H₁₃BrO₂
MW: 317.2 g/mol
Melting Pt: 104…106 °C
Storage Temperature: Freezer
MDL Number: MFCD00270871
CAS Number: 88070-98-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5719f7ff05753fdd343bb90d39435d46946ad6d1&vid=30522af9c9d5c8db61540bb88b91931558454548&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR