2-Methoxyestradiol ≥95%

Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BMLS5400010 BMLS5400050
ENZOBMLS5400010EA 87 GBP
ENZOBMLS5400010 ENZOBMLS5400050
2-Methoxyestradiol ≥95%
2-Methoxyestradiol

Estrogen receptor inhibitor


An endogenous estrogen metabolite with low affinity for the estrogen receptor but which disrupts microtubule function. Potent inhibitor of endothelial cell proliferation and migration. Inhibits bFGF and VEGF-induced corneal neovascularization in mice. Posttranscriptionally downregulates HIF-1alpha expression.

Formula: C₁₉H₂₆O₃
MDL Number: MFCD00010489
CAS Number: 362-07-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=04d88b2dab2bd16909939528bf1f4cd9288ba066&vid=5a0aac6c88da26b90501143473f12468b80a3263&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR