Z-Leu-Arg-Gly-Gly-Amc
Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BMLP8010005
ENZOBMLP8010005EA 250 GBP
ENZOBMLP8010005
Z-Leu-Arg-Gly-Gly-Amc
Proteins and Peptides
Substrate for ubiquitin C-terminal hydrolases (UCHs)
Fluorogenic substrate for ubiquitin C-terminal hydrolases (UCHs), including UCH-L3 and Isopeptidase T. Sequence is based on the C-terminal residues of ubiquitin. Ex.: 380 nm Em.: 460 nm.