Hsp27tide
Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BMLP2460001
ENZOBMLP2460001EA 162 GBP
ENZOBMLP2460001
Hsp27tide
Proteins and Peptides
MAPKAPK substrate
This peptide, RRLNRQLSVA, is derived from Hsp27 (aa 80-85) and is a substrate for MAPKAPK2 and MAPKAPK3.