MMP Substrate (chromogenic)
Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BMLP1250005
ENZOBMLP1250005EA 256 GBP
ENZOBMLP1250005
MMP Substrate (chromogenic)
Proteins and Peptides
Sensitive chromogenic MMP substrate
Chromogenic substrate for continuous spectrophotometric assay of most matrix metalloproteinases, and TACE (ADAM17). The MMP cleavage site peptide bond is replaced by a thioester bond in this peptide. Hydrolysis of this bond by an MMP produces a sulfhydryl group, which reacts with DTNB [5,5'-dithiobis(2-nitrobenzoic acid), Ellman's Reagent] to form 2-nitro-5-thiobenzoic acid, which can be detected by its absorbance at 412 nm.