Phosphatidic acid, dipalmitoyl ≥98% (by TLC), white solid

Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BMLLP1030100
ENZOBMLLP1030100EA 166 GBP
ENZOBMLLP1030100
Phosphatidic acid, dipalmitoyl ≥98% (by TLC), white solid
Phosphatidic acid, dipalmitoyl

PKC activator


A mitogenic lipid phosphate produced via the action of phospholipase D. Mediates agonist-dependent Raf-1 translocation and activation of the MAP kinase cascade.


Fine suspension in ethanol (5 mg/ml) with 15 minutes of sonication at room temperature.

Formula: C₃₅H₆₈O₈P·Na
Storage Temperature: Freezer
CAS Number: 169051-60-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=366b423743368caa6265b5a8118881aa6c5339ee&vid=1b308a259207952ee02088c1d869fabf6aa8ada4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (TLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR