Actinomycin D (from Streptomyces parvulus) ≥98% (by TLC)

Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

BMLGR3000005
ENZOBMLGR3000005EA 87 GBP
ENZOBMLGR3000005
Actinomycin D (from Streptomyces parvulus) ≥98% (by TLC)
Actinomycin D

An intercalating transcription inhibitor and antineoplastic antibiotic. Inhibits transcription by binding to DNA at dG residues. Induces apoptosis in many cell lines, but suppresses in PC12 cells induced by topoisomrease II inhibitor etopside. Used at 5-10 µg/mL in cell culture for halting transcription. The product is not sterile.


Red-orange crystalline solid


Soluble in DMSO (>20 mg/mL), 100% ethanol (1 mg/mL) or methanol

Formula: C₆₂H₈₆N₁₂O₁₆
Storage Temperature: Freezer
MDL Number: MFCD00005033
CAS Number: 50-76-0
UN: 3462
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1cde9b7d28f0132d28e65c4135b94a831303eb34&vid=d38d0403aca86c9ce2901f80c447277fbdf95b76&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR