Troglitazone ≥98% (by TLC)

Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BMLGR2100005 BMLGR2100025
ENZOBMLGR2100005EA 166 GBP
ENZOBMLGR2100005 ENZOBMLGR2100025
Troglitazone ≥98% (by TLC)
Troglitazone

PPAR ligand


Antidiabetic thiazolidinedione. Potent and selective PPARγ ligand. Binds to the PPARγ ligand-binding domain and recruits ligand-specific transcriptional coactivators at 1µM. In skeletal muscle and adipocyte cell cultures, the EC50 of troglitazone is about 10µM. Displaces [125I]SB-236636 with an IC50=7.9 µM. Stimulates adipogenesis and decreases osteoblastogenesis in human mesenchymal stem cells. Induces apoptosis in human vascular smooth muscle cells and inhibits the growth of a variety of human cancer cells including lung, bladder and myeloid leukemia cells.


Synthetic.

Formula: C₂₄H₂₇NO₅S
MDL Number: MFCD00878416
CAS Number: 97322-87-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=10dcf0bfa8bcd161c397ed22b71d3b3e7a351c16&vid=0202869628fd830de451b310c4f15fdecb6885b8&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR