Clostridium difficile toxoid B
Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BMLG1550050
ENZOBMLG1550050EA 1160 GBP
ENZOBMLG1550050
Clostridium difficile toxoid B
Microbiology Test Systems Antigen-Antibody Tests Ag-AK, Seren
Clostridium difficile toxoid A from Clostridium difficile, strain VPI10463 is a high potency toxin applicable to ELISA, antibody capture, cytotoxicity studies, immunogen and LINE assay.
  • Minimal lot-to-lot variation
  • Inactivated by formaldehyde
  • Lyophilised for ease of handling and long term stability