Linoleic acid ≥99%, colourless oil

Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: (9Z,12Z)-Octadeca-9,12-dienoic acid, cis,cis-9,12-Octadecadienoic acid, 9Z,12Z-Octadecadienoic acid

BMLFA0141000
ENZOBMLFA0141000EA 66.1 GBP
ENZOBMLFA0141000
Linoleic acid ≥99%, colourless oil
Linoleic acid

PPAR ligand.


Induces adipocyte differentiation and activates PPARs in cultured cells.


Soluble in DMSO and ethanol.

Formula: C₁₈H₃₂O₂
MW: 280.45 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00064241
CAS Number: 60-33-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc67abdb9ae0460d8cc5d113085a6c1fb3930414&vid=350f40547b15542ca4b5e542eea81a537d01da85&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR