HNMPA-(AM)3 ≥98% (by TLC)

Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Hydroxy-2-naphthalenylmethylphosphonic acid tri-acetoxymethyl ester

BMLEI2480005 BMLEI2480025
ENZOBMLEI2480005EA 177 GBP
ENZOBMLEI2480005 ENZOBMLEI2480025
HNMPA-(AM)3 ≥98% (by TLC)
HNMPA-(AM)3

HNMPA-(AM)3 is a cell-permeable analog of HNMPA. It inhibits insulin receptor tyrosine kinase activity (IC50=100 µM) and insulin-stimulated glucose oxidation in isolated rat adipocytes (IC50=10 µM). It has no effect on PKA (at concs. up to 1 mM) or PKC (at concs. up to 420 µM). Inhibits both serine and tyrosine autophosphorylation by the human insulin receptor. Does not affect protein kinase C (PKC) or cAMP-dependent protein kinase (PKA).


  • Insulin receptor inhibitor


Colorless oil.


Soluble in DMSO (>20mg/ml) or 100% ethanol (>20mg/ml).

Formula: C₂₀H₂₃O₁₀P
Storage Temperature: Freezer
CAS Number: 120944-03-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a98026d7222076fc93ed9689ae88d137a973f81&vid=71f0a0db86a6d3be376ce057451548e88a467199&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR