kainic acid ≥98% (by HPLC), white to off-white solid

Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BMLEA1230005 BMLEA1230025
ENZOBMLEA1230005EA 89.2 GBP
ENZOBMLEA1230005 ENZOBMLEA1230025
kainic acid ≥98% (by HPLC), white to off-white solid
kainic acid

Selective agonist at kainate receptors.


Soluble in water (25 mg/ml).

Formula: C₁₀H₁₅NO₄
Storage Temperature: Ambient
MDL Number: MFCD00077806
CAS Number: 487-79-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc50b335d9ca52ba9c488755b54dd9e6d29df449&vid=8d74394820efe0c9e3d85e351f801565bd8c267c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (HPLC)
Identity Determined by H-NMR


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR