AMG-9810 ≥98%

Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: (E)-3-(4-(tert-butyl)phenyl)-N-(2,3-dihydrobenzo[b][1,4]dioxin-6-yl)acrylamide, AMG9810, (E)-3-(4-(tert-Butyl)phenyl)-N-(2,3-dihydrobenzo[b][1,4]dioxin-6-yl)acrylamide, AMG 9810, (2E)-N-(2,3-Dihydro-1,4-benzodioxin-6-yl)-3-[4-(1,1-dimethylethyl)phenyl]-2-propenamide, 2E-N-(2,3-Dihydro-1,4-benzodioxin-6-yl)-3-[4-(1,1-dimethylethyl)phenyl]-2-Propenamide

BMLCA2320010 BMLCA2320050
ENZOBMLCA2320010EA 111 GBP
ENZOBMLCA2320010 ENZOBMLCA2320050
AMG-9810 ≥98%
AMG-9810

TRPV1 antagonist.


AMG-9810 is an antagonist of capsaicin activation of TRPV1 (IC₅₀ = 24.5 nM) as well as activation by acid, heat, and endogenous ligands.


Soluble in DMSO (>25 mg/ml), ethanol (20 mg/ml).

Formula: C₂₁H₂₃NO₃
MDL Number: MFCD00955103
CAS Number: 545395-94-6
New Product

Order Now

Specification Test Results

Purity ≥98%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR