Hepatotoxicity library, Screen-Well®
Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BML28510100 BML28510500
ENZOBML28510100EA 4010 GBP
ENZOBML28510100 ENZOBML28510500
Hepatotoxicity library, Screen-Well®
Cell Culture Supplements and Additives
The Screen-Well® hepatotoxicity library is a focused collection of 238 compounds with defined and diverse hepatotoxicity, including steatosis, mitochondrial toxicity, Mallory body formation, cholestatic effects, and many more. A variety of structurally and mechanistically different compound classes are included, as well as nontoxic controls. The compounds are dissolved in DMSO at 10 mM and aliquoted into deep-well plates. The library is an essential tool for predictive toxicology screening and assay development.
Ordering information: Contains 238 compounds with defined and diverse hepatotoxicity. A variety of structurally and mechanistically different compound classes, as well as nontoxic controls, are included.