Autophagy library, Screen-Well®
Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BML28370100 BML28370500
ENZOBML28370100EA 1270 GBP
ENZOBML28370100 ENZOBML28370500
Autophagy library, Screen-Well®
Cell Culture Supplements and Additives
Structurally and mechanistically different compounds for studying the roles of pro- and anti-autophagic molecules in cells and in vitro applications.
The Screen-well® autophagy library contains 94 compounds with defined autophagy-inducing or -inhibitory activity. Compounds are dissolved in DMSO at 10 mM or 1 mM and aliquoted into deep-well plates at 100 or 500 µl per well. A variety of structurally and mechanistically different compound classes are included. A useful tool for studying the roles of pro- and anti-autophagic molecules in cells as well as for use in in vitro applications.
Ordering information: Contains 94 compounds with defined autophagy-inducing or -inhibitory activity.  A variety of structurally and mechanistically different compound classes are included.