Protease inhibitor library, Screen-Well®
Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BML28330100 BML28330500
ENZOBML28330100EA 3010 GBP
ENZOBML28330100 ENZOBML28330500
Protease inhibitor library, Screen-Well®
Cell Culture Supplements and Additives
The Screen-Well™ Protease Inhibitor Library includes 53 known protease inhibitors on a single 96-well plate. Inhibitors are supplied dissolved in DMSO at 10 mM and aliquoted into deep-well plates at either 100 or 500 microliters per well. The library includes inhibitors to a broad range of important proteases and is a convenient and cost-effective way to purchase a panel of protease inhibitors for chemical genomics, assay development, and other applications. It includes inhibitors of: Calpains, TPPII, Neutrophil Elastase, Cathepsins, DPPIV, Caspases, Granzyme B, gamma-Secretase, Furin, MMPs, ACE, Kallikrein, Proteasome, Aminopeptidase B, and many more.
The Screen-Well™Protease Inhibitor Library includes 53 known protease inhibitors on a single 96-well plate. Inhibitors are supplied dissolved in DMSO at 10 mM and aliquoted into deep-well plates at either 100 or 500 microliters per well. The library includes inhibitors to a broad range of important proteases and is a convenient and cost-effective way to purchase a panel of protease inhibitors for chemical genomics, assay development, and other applications. It includes inhibitors of: Calpains, TPPII, Neutrophil Elastase, Cathepsins, DPPIV, Caspases, Granzyme B, gamma-Secretase, Furin, MMPs, ACE, Kallikrein, Proteasome, Aminopeptidase B, and many more.