Cholinergic ligand library, Screen-Well®
Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BML28150100 BML28150500
ENZOBML28150100EA 1330 GBP
ENZOBML28150100 ENZOBML28150500
Cholinergic ligand library, Screen-Well®
Cell Culture Supplements and Additives
The cholinergic ligands plate contains 72 compounds ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs. The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The cholinergic ligands plate is 1 plate of the larger neurotransmitter library which contains over 650 ligands.