Opioid ligand library, Screen-Well®
Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BML28140100 BML28140500
ENZOBML28140100EA 2170 GBP
ENZOBML28140100 ENZOBML28140500
Opioid ligand library, Screen-Well®
Cell Culture Supplements and Additives
The Screen-Well® opioid and sigma ligands plate contains 74 ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs. The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The opioid and sigma ligands plate is 1 plate of the larger Neurotransmitter Library which contains over 650 ligands.