Muscimol ≥98% (by TLC)

Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

ALX550127M005
ENZOALX550127M005EA 170 GBP
ENZOALX550127M005
Muscimol ≥98% (by TLC)
Muscimol

Potent GABAA receptor agonist.


Very soluble in water (10 mg/ml) or methanol; soluble in 100% ethanol.

Formula: C₄H₆N₂O₂
Storage Temperature: Freezer
MDL Number: MFCD00057894
CAS Number: 2763-96-4
EINECS: 220-430-4
UN: 1544
ADR: 6.1,II
Merck Index: 14,06312

Order Now

Specification Test Results

Purity (TLC) ≥98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR