Human Recombinant CD137:Fc
Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ALX522031C050
ENZOALX522031C050EA 588 GBP
ENZOALX522031C050
Human Recombinant CD137:Fc
Proteins and Peptides
Produced in HEK 293 cells. The cysteine-rich region of human CD137 (4-1BB) (aa 1-186) is fused to the Fc portion of human IgG1.