Methyl-6-O-(N-heptylcarbamoyl)-α-D-glucopyranoside ≥98% (by HPLC)

Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ALX500003G001 ALX500003G005
ENZOALX500003G001EA 73.8 GBP
ENZOALX500003G001 ENZOALX500003G005
Methyl-6-O-(N-heptylcarbamoyl)-α-D-glucopyranoside ≥98% (by HPLC)
Methyl-6-O-(N-heptylcarbamoyl)-alpha-D-glucopyranoside

Non-ionic, glucose based very mild detergent for membrane protein purification.


Hecameg is used for the extraction, purification, stabilisation of proteins (recombinant or natural proteins). Surfactant for chromatography, electrophoresis and ELISA analysis. Extraction of other biomolecules (DNA and RNA) from proteous samples. Also used for the study of protein structure; reconstitution or crystallisation of membrane proteins, enzymes or antigens; liposomes preparation. Sanitisation of chromatography columns.


Soluble in water (>100 mg/ml at +4 °C).

Formula: C₁₅H₂₉NO₇
Storage Temperature: Refrigerator
MDL Number: MFCD00077425
CAS Number: 115457-83-5

Order Now

Specification Test Results

Purity (HPLC) ≥98%
Appearance White to off-white crytalline solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR