Leflunomide ≥99%

Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 5-Methyl-N-(4-(trifluoromethyl)phenyl)isoxazole-4-carboxamide, 5-Methyl-N-[4-(trifluoromethyl)phenyl]isoxazole-4-carboxamide

ALX430095G001 ALX430095M050
ENZOALX430095G001EA 769 GBP
ENZOALX430095G001 ENZOALX430095M050
Leflunomide ≥99%
Leflunomide

Immunosuppressant


Immunosuppressive drug that inhibits T and B cell responses in vivo. Inhibitor of tyrosine phosphorylation (tyrosine kinase inhibitor), of dihydroorotate dehydrogenase, an enzyme for de novo pyrimidine synthesis, of TNF-dependent NF-kappaB activation, and of COX-2. The physiological activity of leflunomide is attributed mainly to its metabolite A77 1726 (Prod. No. ALX-430-096).

Formula: C₁₂H₉F₃N₂O₂
MW: 270.21 g/mol
MDL Number: MFCD00867593
CAS Number: 75706-12-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR