Doxycycline hyclate ≥98% (dried basis)

Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Doxycycline Hydrochloride Hemiethanolate Hemihydrate

ALX380273G001 ALX380273G005
ENZOALX380273G001EA 51.8 GBP
ENZOALX380273G001 ENZOALX380273G005
Doxycycline hyclate ≥98% (dried basis)
Doxycycline hyclate

MMP inhibitor


Broad spectrum antibiotic and bacteriostatic. Shows antiprotozoal properties. Potent inhibitor of MMPs (matrix metalloproteinases) in vivo. Inhibits collagen synthesis.


Semisynthetic from oxytetracycline.

Formula: C₂₂H₂₄N₂O₈·HCl·0.5H₂O·0.5C₂H₆O
MDL Number: MFCD07357237
CAS Number: 24390-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR