Olomoucine ≥98%, high purity

Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 2-((6-(Benzylamino)-9-methyl-9H-purin-2-yl)amino)ethanol

ALX350013M025
ENZOALX350013M025EA 469 GBP
ENZOALX350013M025
Olomoucine ≥98%, high purity
Olomoucine

Olomoucine is a purine derivative which acts as a potent and selective inhibitor of CDK1 (p34cdc2)/cyclin B and related kinases. It acts by competing for the ATP binding domain of the kinase. Of 35 protein kinases tested, olomoucine inhibited p33CDK2/cyclin A, p33CDK2/cyclin E, p34cdc2/cyclin B, p33CDK5/p35 and erk1 with IC50 of 7, 7, 7, 3 and 25µM respectively. The related kinases p34CDK4/cyclin D and p40CDK6/cyclin D3 were not inhibited or were less sensitive to olomoucine, IC50 > 1000 and 250µM respectively. PKC, PKA, PKG, CaM kinase II, MLC kinase, CK2, EGFR tyrosine kinase, p56lck, p56lyn, p60c-src, v-abl and insulin receptor tyrosine kinase are not inhibited or are weakly inhibited at 1mM. It delays mouse oocyte maturation and inhibits the growth of a variety of human cell lines including Rhabdomyosarcoma cell line Rh1, Rh18, Rh28 and Rh30; MCF-7, KB-3-1 and their adriamycin-resistant counterparts (IC50=18-86µM). It inhibits DNA synthesis in interleukin-2-stimulated T lymphocytes and triggers a G1 arrest similar to interleukin-2 deprivation. Can be used to synchronize cells in G1. Triggers apoptosis in target tumor cells in vitro and in vivo and in maturing cerebellar granule neurons. N9-Isopropyl-olomoucine and the negative control Iso-olomoucine are also available.


  • CDK/cyclin inhibitor

Formula: C₁₅H₁₈N₆O
MDL Number: MFCD00189360
CAS Number: 101622-51-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR