Tannic acid 95

Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Gallotannin, Tannin

ALX270418G001
ENZOALX270418G001EA 50.7 GBP
ENZOALX270418G001
Tannic acid 95
Tannic acid

Inhibitor of PARG and eNOS


Inhibitor of poly(ADP-ribose) glycohydrolase (PARG). Cytoprotective in oxidatively stressed cells. Inhibitor of endothelial nitric oxide synthase (eNOS; NOS III) and weak inhibitor of inducible (iNOS; NOS II) and neuronal nitric oxide synthase (nNOS; NOS I). Induces cyclooxygenase-2 (COX-2) expression. Free radical scavenger.


Occurs in the bark and fruit of many plants, notably in the bark of the oak species, in sumac and myrobalan.

Formula: C₇₆H₅₂O₄₆
MW: 1701.2 g/mol
Flash Pt: 199
MDL Number: MFCD00066397
CAS Number: 1401-55-4
Merck Index: 12,09221

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR